Learn More
Biotium BAPTA, tetrapotassium salt
Supplier:  Biotium 50002
Encompass_Preferred
BAPTA and its derivatives are calcium chelators that are commonly used to form calcium buffers with well-defined calcium concentrations. By injecting the chelators into cells or by incubating cells with the AM ester form of the chelators, one can control the cytosolic calcium concentration, an important means to study the roles of calcium. Key advantages of these calcium chelators include relative insensitivity toward intracellular pH change and fast release of calcium. Biotium offers several BAPTA chelators with calcium dissociation constants covering the biologically significant range from 10-7 to 10-2 M. Features: Kd (no Mg2 ): 0.59 uM, Kd (1 mM Mg2 ): 0.70 uM; White solid soluble in water; Store at 4°C; C23H24K4N2O10; MW: 656.85; [73630-08-7]
Catalog No. NC0537551