Learn More
Aat Bioquest BAPTA tetrapotassium salt CAS 73630-08-7 (100 mg)
SDP
Catalog No. 502249900
Encompass_Preferred
BAPTA 12-biso-aminophenoxyethane-NNNN-tetraacetic acid is a water-soluble and cell-impermeable calcium chelator. This catalog is for 100 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9900 Supplier Aat Bioquest Supplier No. 21003
BAPTA 12-biso-aminophenoxyethane-NNNN-tetraacetic acid is a water-soluble and cell-impermeable calcium chelator. This catalog is for 100 mg.