Learn More
Aviva Systems Biology Corp Bapx1 Antibody - N-terminal region (ARP36826_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36826_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bapx1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse Bapx1. Alternate names: NKX3.2, Nkx-3.2, Nkx3-2, Bapx1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53058