Learn More
Grainger DEWALT Trigger Clamp, Steel, 6 in., 2/Pk.
Catalog No. 502778493
Encompass_Preferred
These 6 in. medium-duty bar clamps are designed for versatile, one-handed operation. Featuring reinforced nylon bodies and durable heat-treated steel bars, they provide reliable clamping force for a wide range of applications. Each unit includes removable jaw pads to help minimize marring during use, and the integrated quick-change button allows for easy conversion to a spreader.
  • 6 in. maximum clamping capacity
  • 4 in. to 10 in. spreading range
  • 100 lb. clamping force
  • Removable, grooved jaw pads included
  • One-handed trigger operation
  • Set of 2 clamps
Catalog No. 50-277-8493 Supplier Grainger Supplier No. 18G796
These 6 in. medium-duty bar clamps are designed for versatile, one-handed operation. Featuring reinforced nylon bodies and durable heat-treated steel bars, they provide reliable clamping force for a wide range of applications. Each unit includes removable jaw pads to help minimize marring during use, and the integrated quick-change button allows for easy conversion to a spreader.
  • 6 in. maximum clamping capacity
  • 4 in. to 10 in. spreading range
  • 100 lb. clamping force
  • Removable, grooved jaw pads included
  • One-handed trigger operation
  • Set of 2 clamps