Learn More
Electron Microscopy Sciences Veco Parallel Bar with Divider R100Aspec, Copper
SDP
Catalog No. 5027653
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm
Catalog No. 50-276-53 Supplier Electron Microscopy Sciences Supplier No. R100ASCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm