Learn More
Med Vet International Bar-Vac CD/T Cattle, Sheep & Goat Vaccine, 50mL, Each
Catalog No. 502828816
Encompass_Preferred
Bar-Vac CD/T is recommended for the immunization of healthy sheep, goats and cattle against enterotoxemia and tetanus caused by the toxins of Clostridium perfringens types C & D (overeating disease) and Clostridium tetani (tetanus.) Dosage is 2 ml for sheep and goats and 5 ml for cattle injected subcutaneously. Repeat in 21 to 28 days and an annual booster of Bar-Vac is recommended.
Catalog No. 50-282-8816 Supplier Med Vet International Supplier No. DTITOXIN50
Bar-Vac CD/T is recommended for the immunization of healthy sheep, goats and cattle against enterotoxemia and tetanus caused by the toxins of Clostridium perfringens types C & D (overeating disease) and Clostridium tetani (tetanus.) Dosage is 2 ml for sheep and goats and 5 ml for cattle injected subcutaneously. Repeat in 21 to 28 days and an annual booster of Bar-Vac is recommended.