Learn More
Medchemexpress LLC Bar501 | 1632118-69-4 | MFCD30738007 | 99.9% | 406.64 | C26H46O3 | 5 MG
SDP
Catalog No. 5000212750
Encompass_Preferred
BAR501 is a cholane-scaffold bile acid derivative and a selective agonist of the GPBAR1 (TGR5) receptor, provided for research use. It has well-defined physicochemical properties and documented solubility information to support in vitro assays and in vivo formulation development.
  • Selective GPBAR1 (TGR5) agonist useful for receptor pharmacology studies.
  • High purity (99.90%) suitable for research applications.
  • Defined molecular formula and weight for analytical reproducibility.
  • Available in small pack sizes suitable for exploratory studies.
  • Documented in vitro solubility in ethanol and DMSO for assay preparation.
  • In vivo formulation guidance provided to support preclinical dosing.
Catalog No. 50-002-12750 Supplier Medchemexpress LLC Supplier No. HY101274A5MG
BAR501 is a cholane-scaffold bile acid derivative and a selective agonist of the GPBAR1 (TGR5) receptor, provided for research use. It has well-defined physicochemical properties and documented solubility information to support in vitro assays and in vivo formulation development.
  • Selective GPBAR1 (TGR5) agonist useful for receptor pharmacology studies.
  • High purity (99.90%) suitable for research applications.
  • Defined molecular formula and weight for analytical reproducibility.
  • Available in small pack sizes suitable for exploratory studies.
  • Documented in vitro solubility in ethanol and DMSO for assay preparation.
  • In vivo formulation guidance provided to support preclinical dosing.