Learn More
Cayman Chemical BAR501 ImpurIty 10mg
SDP
Catalog No. 502964127
Encompass_Preferred
An impurity found in the preparation of BAR501; GP-BAR1 agonist; induces a 150% increase in luciferase activity in a GP-BAR1 reporter gene assay at a concentration of 10 µM
Catalog No. 50-296-4127 Supplier Cayman Chemical Supplier No. 2246110 MG
An impurity found in the preparation of BAR501; GP-BAR1 agonist; induces a 150% increase in luciferase activity in a GP-BAR1 reporter gene assay at a concentration of 10 µM