Learn More
Selleck Chemical LLC BAR502 S0033-25MG
SDP
Catalog No. 5000039722
Encompass_Preferred
BAR502 (Compound 30) is a dual FXR and GPBAR1 agonist with IC50 values of 2 M and 0 4 M respectively
Catalog No. 50-000-39722 Supplier Selleck Chemical LLC Supplier No. S003325MG
BAR502 (Compound 30) is a dual FXR and GPBAR1 agonist with IC50 values of 2 M and 0 4 M respectively