Learn More
Medchemexpress LLC HY-101273 5mg Medchemexpress, BAR502 CAS:1612191-86-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012735MG
Encompass_Preferred
Medchemexpress, HY-101273 5mg BAR502 CAS:1612191-86-2 BAR502 is a dual FXR and GPBAR1 agonist with IC 50 values of 2 μM and 0.4 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2083