Learn More
Selleck Chemical LLC Barasertib (AZD1152-HQPA) S1147-25mg
SDP
Catalog No. 503108157
Encompass_Preferred
Defosbarasertib (AZD1152-HQPA AZD2811 INH-34 Barasertib-HQPA) is a highly selective Aurora B inhibitor with IC50 of 0 37 nM in a cell-free assay 3700 fold more selective for Aurora B over Aurora A Phase 1
Catalog No. 50-310-8157 Supplier Selleck Chemical LLC Supplier No. S114725MG
Defosbarasertib (AZD1152-HQPA AZD2811 INH-34 Barasertib-HQPA) is a highly selective Aurora B inhibitor with IC50 of 0 37 nM in a cell-free assay 3700 fold more selective for Aurora B over Aurora A Phase 1