Learn More
Selleck Chemical LLC Barasertib (AZD1152-HQPA) 5mg 722544-51-6 AZD2811
SDP
Catalog No. 507802
Encompass_Preferred
Barasertib (AZD1152-HQPA, AZD2811) is a highly selective Aurora B inhibitor with IC50 of 0.37 nM in a cell-free assay, ~3700 fold more selective for Aurora B over Aurora A. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-780-2 Supplier Selleck Chemical LLC Supplier No. S11475MG
Barasertib (AZD1152-HQPA, AZD2811) is a highly selective Aurora B inhibitor with IC50 of 0.37 nM in a cell-free assay, ~3700 fold more selective for Aurora B over Aurora A. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.