Learn More
AVANTOR MASTERFLEX (VWR LLC) NPT Male Adapter to Hose Barb, 1/2"" NPT to 1/2"" ID, PP, 10/Pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4061046/DEL
Encompass
NPT Male Adapter to Hose Barb, 1/2" NPT to 1/2" ID, Polypropylene, 10/Pk, Sterilization Method- Ethylene oxide (ETO) or gamma
Catalog No. NC1032943