Learn More
Cole-Parmer Hose Barb Fitting, Straight Union, HDPE, Cleanroom Packed, 3/8" ID; 10/Pk
Supplier:  Cole-Parmer 5010839
Encompass_Preferred
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
Catalog No. 50-255-8388