Learn More
Cole-Parmer Hose Barb to Plug Fitting, Straight Adapter, Natural Kynar, Cleanroom Packed, 3/4" ID; 10/Pk
Catalog No. 502558163 Shop All Cole-Parmer Products
Encompass_Preferred
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Simply insert the hose barb plug in the soft tubing to stop the fluid flow. A single barb on one end of the fitting creates a tight seal to keep the tubing in place. These natural Kynar fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
Catalog No. 50-255-8163 Supplier Cole-Parmer Supplier No. 5011064
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Simply insert the hose barb plug in the soft tubing to stop the fluid flow. A single barb on one end of the fitting creates a tight seal to keep the tubing in place. These natural Kynar fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.