Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Sanitary Clamp to Hose Barb Adapter with TPE Fused Gasket, 1 1/2"" Clamp x 3/8"" ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3120134
Encompass
Sanitary fitting with built-in gasket makes sanitary installation easier than ever while adding another layer of safety to installation. No user contact with the gasket ensures sterility and prevents contamination. The limited slip design eliminates the risk of misalignment.
Catalog No. NC1676543