Learn More
Analytical Sales & Services Barbed Adapter for Waste Manifold Outlet Port, for 6-9mm ID Tubing, each
Catalog No. NC3793221
Encompass
Barbed Adapter for Waste Manifold Outlet Port, for 6-9mm ID TubingQty/Pkg: EACH
Catalog No. NC3793221 Supplier Analytical Sales & Services Supplier No. BA0069
Barbed Adapter for Waste Manifold Outlet Port, for 6-9mm ID TubingQty/Pkg: EACH