Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, 303 Stainless Steel, Tee, Hose Barb Reducer, 3/8"" ID x 3/8"" ID x 1/8"" ID
Catalog No. NC1504773
Encompass
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. NC1504773 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3120847
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).