Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Black Polypropylene, Cross Connector, Hose Barb, 1/4""; 5/PK
Catalog No. NC1482014
Encompass
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. Cross connectors feature four hose barb ports.
Catalog No. NC1482014 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140711
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. Cross connectors feature four hose barb ports.