Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Natural Polypropylene, Straight, Hose Barb Reducer, 1/2"" ID x 1/4"" ID; 100/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4064010/DEL
Encompass
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. NC1486098