Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Clear Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/8"" ID x 1/4""-28 UNF(M); 25/PK
Catalog No. NC0369760
Encompass
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. NC0369760 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3150154
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).