Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Sanitary Clamp to Hose Barb Adapter, 3/4"" Clamp x 1/4"" ID
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3180525/DEL
Encompass
Allowing for easy assembly and disassembly of sanitary set-ups, these adapters are suitable for use connecting a Tri-Clamp fitting to flexible tubing with the hose barb adapter. Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.
Catalog No. NC9851111