Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, 316 Stainless Steel, Straight, Hose Barb to Threaded Adapter, 3/8"" ID x 3/8"" NPT(M)
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3120886
Encompass
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
Catalog No. NC1527539