Learn More
Waters Corp Barbed Straight Connector, 1/4 in. ID, Nylon
Catalog No. 502390473 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This straight connector is a replacement part designed for use on Arc systems, primarily for routing solvent waste. It features a 1/4 in. ID and is constructed from durable nylon.
  • Designed for Arc systems
  • Ideal for routing solvent waste
  • Features a 1/4 in. ID
  • Constructed from durable nylon
  • Functions as a fitting
  • Compatible with UPLC systems
  • Packaged individually
  • Replacement as needed
Catalog No. 50-239-0473 Supplier Waters Corp Supplier No. 700010705
This straight connector is a replacement part designed for use on Arc systems, primarily for routing solvent waste. It features a 1/4 in. ID and is constructed from durable nylon.
  • Designed for Arc systems
  • Ideal for routing solvent waste
  • Features a 1/4 in. ID
  • Constructed from durable nylon
  • Functions as a fitting
  • Compatible with UPLC systems
  • Packaged individually
  • Replacement as needed