Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barbituric Acid, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608954 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
Packaging:
2500cc White opaque plastic container
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-954 25 g Poly Bottle
18-608-953 2.5 kg 2500cc White opaque plastic container
18-608-952 125 g Poly Bottle
18-608-956 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-954 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BA12025GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-52-7

Specifications

CAS Max % 1
Quantity 25 g
Packaging Poly Bottle
Percent Purity 99%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.