Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, PVDF, 5/8 x 7/8 in
Supplier:  Accuflow Systems BL135625ALPVDF
Encompass_Preferred
One-piece PVDF tubing retainer that secures barbed tubing connections with a full 360° radial crimp and compression over the barb ridge to minimize leaks. The single-piece sleeve-and-collet design reduces assembly steps and helps prevent assembly errors, while PVDF construction provides chemical resistance and biocompatibility for critical fluid transfer applications.
  • Secures barbed tubing connections with a full 360° radial crimp.
  • Provides 360° compression directly over the barb ridge for leak resistance.
  • Manufactured from PVDF for chemical resistance and biocompatibility.
  • Compatible with a wide range of flexible tubing materials.
  • Single-piece design simplifies assembly and reduces assembly errors.
  • Sized for 5/8 in barb to 7/8 in tube outer diameter connections.
Catalog No. NC2827614