Learn More
Accuflow Systems BarbLock Tubing Retainer
Catalog No. NC3936007
Encompass_Preferred
This BarbLock tubing retainer is a one-piece clamp designed for securing tubing made from polypropylene, ensuring a reliable connection for various applications.
  • Made from polypropylene material
  • One-piece configuration for secure connections
  • Designed for 0.5000 in. (12.7 mm) inner diameter tubing
  • Designed for 0.7500 in. (19.05 mm) outer diameter tubing
  • Compatible with CPC and PendoTech systems
  • Coloring agent biocompatibility tested to ISO, USP Class VI standards
Catalog No. NC3936007 Supplier Accuflow Systems Supplier No. BL135506ALLRR
This BarbLock tubing retainer is a one-piece clamp designed for securing tubing made from polypropylene, ensuring a reliable connection for various applications.
  • Made from polypropylene material
  • One-piece configuration for secure connections
  • Designed for 0.5000 in. (12.7 mm) inner diameter tubing
  • Designed for 0.7500 in. (19.05 mm) outer diameter tubing
  • Compatible with CPC and PendoTech systems
  • Coloring agent biocompatibility tested to ISO, USP Class VI standards