Learn More
LumiraDx Barcode Scanner
Catalog No. 22181004 Shop All Roche Diagnostics Products
Change view
Click to view available options
Type:
Barcode Scanner
Catalog No. Type
22-181-004 Barcode Scanner
1 options
Catalog No. 22-181-004 Supplier LumiraDx Supplier No. 10417927001
Add to Cart
Add to Cart
LumiraDx barcode scanner for use with the LumiraDx Platform
  • Features:
    • For use with the LumiraDx platform
    • Capable of 1-d and 2-d barcodes to improve testing workflow
    • Reduces errors in specimen ID entry and supports operator login or badge access
TRUSTED_SUSTAINABILITY
Specifications
Type Barcode Scanner