Learn More
Mettler Toledo™ Barcode Set SQKitReader
Catalog No. 01911520 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Barcode Scanner
Catalog No. Type
01-911-520 Barcode Scanner
1 options
This item is not returnable. View return policy
Catalog No. 01-911-520 Supplier Mettler Toledo™ Supplier No. 30705183
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mettler Toledo SQKitReader or ScanStart function scans Spectroquant™ water test kit barcodes to automatically start methods inputted on the user-friendly touchscreen.
  • Mettler Toledo Spectrophotometers utilize Spectroquant™ water test kits to calculate the amount of light absorbed by the water and reagent during the test.
  • The SQKitReader increases efficiency by scanning a barcode on the water test kit, so it can automatically begin a method for the user.
TRUSTED_SUSTAINABILITY
Specifications
Type Barcode Scanner
Voltage 120 V