Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Bar-coded Cryogenic Vials

Catalog No. 07200960 Shop All Corning Products
Change view
Click to view available options
Barcode:
1D
2D
Thread Style:
External
External threaded
Internal
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Barcode Thread Style
07-200-960 2D Internal
07-200-961 2D External threaded
07-200-962 1D Internal
07-200-963 1D External
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-200-960 Supplier Corning™ Supplier No. 8670
Add to Cart
Add to Cart
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.

  • Manufactured from polypropylene
  • Leak proof seal for integrity of storage
  • Autoclavable vials and caps, can be re-sterilized
  • Unique 1D, 2D bar-codes and human readable characters
  • Features star shaped cavity in cap and is compatible with decapping equipment
  • Locking base stabilizes tubes for automated handling
  • Graduations and writing patch for manual use also
  • Each tube is checked for clarity and readability of 2D code
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Orange
Color Natural
Sterility Sterile
Volume (Metric) 2 mL
For Use With (Application) Cell biology, cell banking, compound cryopreservation
No. per Case 500
No. per Pack 50
Thread Style Internal
Base Style Round, Self-standing
Barcode 2D
Closure Material Polyethylene
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.