Learn More
Corning™ Bar-coded Cryogenic Vials
Catalog No. 07200961 Shop All Corning Products
Change view
Click to view available options
Barcode:
1D
2D
Thread Style:
External
External threaded
Internal
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Barcode Thread Style
07-200-961 2D External threaded
07-200-960 2D Internal
07-200-962 1D Internal
07-200-963 1D External
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-200-961 Supplier Corning™ Supplier No. 8671
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.
  • Manufactured from polypropylene
  • Leak proof seal for integrity of storage
  • Autoclavable vials and caps, can be re-sterilized
  • Unique 1D, 2D bar-codes and human readable characters
  • Features star shaped cavity in cap and is compatible with decapping equipment
  • Locking base stabilizes tubes for automated handling
  • Graduations and writing patch for manual use also
  • Each tube is checked for clarity and readability of 2D code
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 2 mL
For Use With (Application) Cell biology, cell banking, compound cryopreservation
Thread Style External threaded
Barcode 2D
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov