Learn More
Aspen Surgical™ Bard-Parker™ Protected Disposable Scalpel
Catalog No. 0268880
Change view
Click to view available options
Size:
# 10
# 11
# 15
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
02-688-80 # 15
02-688-78 # 10
02-688-79 # 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 02-688-80 Supplier Aspen Surgical™ Supplier No. 372615
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Features one-handed, locking retractable shield
  • Designed to minimize the risk of surgical blade injuries
  • Features a BD Bard-Parker stainless-steel blade, nonslip grip, and a centimeter scale on the plastic, surgical-green handle
  • Clear protective shield locks both when the blade is covered for protection before and after use, and when blade is uncovered for security and control during use
  • Top button provides tactile sensation as well as an audible click to confirm lock; allows for left or right hand activation
  • Indicator line offers visual confirmation of lock
  • Autoclavable for procedural kits and on-site sterilization
Specifications
Blade Material Stainless Steel
Product Type Safety Scalpel
For Use With (Application) Tissue Separation and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
Size # 15
Disposable Single-use
Sterility Sterile
Certifications/Compliance OSHA
Autoclavable Autoclavable