Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blade Handles
Catalog No. 089154
Change view
Click to view available options
Size:
# 3
# 4
# 7
# 9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
08-915-4 # 4
08-915-2 # 3
08-915-6 # 7
08-915-7 # 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 08-915-4 Supplier Aspen Surgical™ Supplier No. 371040
Add to Cart
Add to Cart
  • Handles are designed for a contoured, firm grip and easy manipulation
  • Distal ends are tapered and beveled for blunt dissection use
Specifications
Blade Material Carbon Steel
Product Type Contoured Blade
Handle Type Surgical Scalpel Handle
Size # 4
For Use With (Equipment) # 20 to # 24 Blade for General Surgery like Tissue Separation and other procedures that require a Sharp Surgical Blade to Puncture or Cut
Disposable Reusable
Sterility Non-sterile