Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Catalog No. 0891510
Change view
Click to view available options
Size:
# 10
# 11
# 15
# 20
# 21
# 22
# 23
# 25
Sterility:
Non-sterile
Sterile
Handle Type:
Ribbed
Safetylock Ribbed
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sterility Handle Type
08-915-10 # 20 Sterile Ribbed
08-915-11 # 21 Sterile Ribbed
08-914C # 20 Non-sterile Ribbed
08-914A # 10 Non-sterile Ribbed
08-914B # 11 Non-sterile Ribbed
08-915-1A # 15 Non-sterile Ribbed
08-914D # 21 Non-sterile Ribbed
08-914E # 23 Non-sterile Ribbed
08-915-13 # 11 Sterile Safetylock Ribbed
08-915-14 # 15 Sterile Safetylock Ribbed
08-915-1B # 22 Non-sterile Ribbed
08-915-1C # 25 Non-sterile Ribbed
08-915-17 # 22 Sterile Safetylock Ribbed
08-915-16 # 21 Sterile Safetylock Ribbed
08-915-9 # 15 Sterile Ribbed
08-915-8 # 10 Sterile Ribbed
22-027982 # 22 Sterile Ribbed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 08-915-10 Supplier Aspen Surgical™ Supplier No. 371120
Only null left
Add to Cart
Add to Cart
Offer consistent sharpness, control and strength
  • Each blade packaged in foil pouch with liner for maximum protection of blade and user
  • Rib back blade design on select blades offers greater cutting control and strength
  • Available with plastic shield SafetyLock to securely cover the blade edge before and after use
  • Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
Blade Material Carbon Steel
Product Type Surgical Blade
For Use With (Application) For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
Handle Type Ribbed
Size # 20
Sterility Sterile