Learn More
Medchemexpress LLC HY-14909 10mg Medchemexpress, Bardoxolone CAS:218600-44-3 Purity:>98%
SDP
Catalog No. 502029076
Encompass_Preferred
Medchemexpress, HY-14909 10mg Bardoxolone CAS:218600-44-3 Bardoxolone is a novel nuclear regulator factor (Nrf-2) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9076 Supplier Medchemexpress LLC Supplier No. HY1490910MG
Medchemexpress, HY-14909 10mg Bardoxolone CAS:218600-44-3 Bardoxolone is a novel nuclear regulator factor (Nrf-2) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.