Learn More
Selleck Chemical LLC Bardoxolone Methyl 25mg 218600-53-4 RTA 402, TP-155, NSC 713200, CDDO Methyl Ester, CDDO-Me
SDP
Supplier:  Selleck Chemical LLC S807825MG
Encompass_Preferred
Bardoxolone Methyl (RTA 402, TP-155, NSC 713200, CDDO Methyl Ester, CDDO-Me) is an IKK inhibitor, showing potent proapoptotic and anti-inflammatory activities; Also a potent Nrf2 activator and nuclear factor-?B (NF-?B) inhibitor. Bardoxolone Methyl abrogates ferroptosis. Bardoxolone methyl induces apoptosis and autophagy in cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0801110