Learn More
Medchemexpress LLC HY-13324 50mg Medchemexpress, Bardoxolone methyl CAS:218600-53-4 Purity:>98%
SDP
Catalog No. 502028983
Encompass_Preferred
Medchemexpress, HY-13324 50mg Bardoxolone methyl CAS:218600-53-4 Bardoxolone methyl (NSC 713200; RTA 402; CDDO Methyl ester) is a synthetic triterpenoid compound with potential antineoplastic and anti-inflammatory activities, acting as an activator of the Nrf2 pathway and an inhibitor of the NF-κB pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8983 Supplier Medchemexpress LLC Supplier No. HY1332450MG
Medchemexpress, HY-13324 50mg Bardoxolone methyl CAS:218600-53-4 Bardoxolone methyl (NSC 713200; RTA 402; CDDO Methyl ester) is a synthetic triterpenoid compound with potential antineoplastic and anti-inflammatory activities, acting as an activator of the Nrf2 pathway and an inhibitor of the NF-κB pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.