Learn More
Medchemexpress LLC HY-13324 200mg , Bardoxolone (methyl) NSC 713200;RTA 402;CDDO Methyl ester CAS:218600-53-4 Purity:98%
SDP
Catalog No. 501492556
Encompass_Preferred
Medchemexpress, HY-13324 200mg Bardoxolone (methyl) NSC 713200;RTA 402;CDDO Methyl ester CAS:218600-53-4 Formula:C32H43NO4 Purity:98% Bardoxolone (methyl) is a potent activator of the nuclear factor erythroid 2-related factor 2 ( Nrf2 ), and uesd for antioxidative and anti-inflammatory treatment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2556 Supplier Medchemexpress LLC Supplier No. HY13324200MG
Medchemexpress, HY-13324 200mg Bardoxolone (methyl) NSC 713200;RTA 402;CDDO Methyl ester CAS:218600-53-4 Formula:C32H43NO4 Purity:98% Bardoxolone (methyl) is a potent activator of the nuclear factor erythroid 2-related factor 2 ( Nrf2 ), and uesd for antioxidative and anti-inflammatory treatment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.