Learn More
Cayman Chemical CDDO, 218600-44-3, 1 mg
SDP
Supplier:  Cayman Chemical 810351
Encompass_Preferred
A synthetic oleanane triterpenoid that blocks iNOS and COX-2 synthesis in INF-γ-activated mouse macrophages (IC50 = 0.4 nM); inhibits proliferation and induces apoptosis in various cancer cell lines and upregulates genes involved in Nrf2/ARE signaling
Catalog No. NC0860060