Learn More
Medchemexpress LLC HY-15315 10mg Medchemexpress, Baricitinib CAS:1187594-09-7 Purity:>98%
SDP
Catalog No. 502029117
Encompass_Preferred
Medchemexpress, HY-15315 10mg Baricitinib CAS:1187594-09-7 Baricitinib (LY3009104; INCB028050) is a selective and orally bioavailable JAK1 and JAK2 inhibitor with IC50s of 5.9 nM and 5.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9117 Supplier Medchemexpress LLC Supplier No. HY1531510MG
Medchemexpress, HY-15315 10mg Baricitinib CAS:1187594-09-7 Baricitinib (LY3009104; INCB028050) is a selective and orally bioavailable JAK1 and JAK2 inhibitor with IC50s of 5.9 nM and 5.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.