Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Baricitinib | 1187594-09-7 | C16H17N7O2S | 250 MG
SDP

Catalog No. 5000374309
Encompass_Preferred

Baricitinib is a selective and orally bioavailable JAK1 and JAK2 inhibitor. It exhibits high purity and is provided as a white to off-white solid, suitable for research applications. Its potent inhibitory activity against JAK1 and JAK2 makes it valuable for studies related to JAK/STAT signaling and epigenetics.

  • Selective JAK1 and JAK2 inhibitor
  • Purity: 99.93%
  • Appearance: Solid, white to off-white
  • Solubility in DMSO: 25 mg/mL (with warming/ultrasonic)
  • JAK1 IC50: 5.9 nM; JAK2 IC50: 5.7 nM
  • Tyk2 IC50: 53 nM; JAK3 IC50: 560 nM
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 1 year, -20°C for 6 months

Catalog No. 50-003-74309 Supplier Medchemexpress LLC Supplier No. 250MGHY15315
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Baricitinib is a selective and orally bioavailable JAK1 and JAK2 inhibitor. It exhibits high purity and is provided as a white to off-white solid, suitable for research applications. Its potent inhibitory activity against JAK1 and JAK2 makes it valuable for studies related to JAK/STAT signaling and epigenetics.

  • Selective JAK1 and JAK2 inhibitor
  • Purity: 99.93%
  • Appearance: Solid, white to off-white
  • Solubility in DMSO: 25 mg/mL (with warming/ultrasonic)
  • JAK1 IC50: 5.9 nM; JAK2 IC50: 5.7 nM
  • Tyk2 IC50: 53 nM; JAK3 IC50: 560 nM
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 1 year, -20°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.