Learn More
Medchemexpress LLC Baricitinib | 1187594-09-7 | MFCD21608464 | 99.9% | 371.42 g/mol | C16H17N7O2S | 10 MG
SDP
Catalog No. 5000285997
Encompass_Preferred
Baricitinib (Standard) is an analytical reference standard of the JAK1/2 inhibitor baricitinib supplied for research and analytical use. Supplied as a 10 mg quantity with reported purity of approximately 99.9%, it includes molecular identification data (C16H17N7O2S, 371.42 g/mol, CAS 1187594-09-7) to support method validation and compound confirmation. Typical applications include calibration, quantitative assays, and method development for chromatographic and mass-spectrometric workflows.
  • High purity (~99.9%) for accurate quantitation.
  • Suitable for HPLC and LC-MS method development.
  • Compact 10 mg quantity for standard preparation and testing.
  • Includes molecular and CAS identification data for traceability.
  • Intended for analytical and research use only.
Catalog No. 50-002-85997 Supplier Medchemexpress LLC Supplier No. HY15315R10MG
Baricitinib (Standard) is an analytical reference standard of the JAK1/2 inhibitor baricitinib supplied for research and analytical use. Supplied as a 10 mg quantity with reported purity of approximately 99.9%, it includes molecular identification data (C16H17N7O2S, 371.42 g/mol, CAS 1187594-09-7) to support method validation and compound confirmation. Typical applications include calibration, quantitative assays, and method development for chromatographic and mass-spectrometric workflows.
  • High purity (~99.9%) for accurate quantitation.
  • Suitable for HPLC and LC-MS method development.
  • Compact 10 mg quantity for standard preparation and testing.
  • Includes molecular and CAS identification data for traceability.
  • Intended for analytical and research use only.