Learn More
Apexbio Technology LLC Baricitinib (LY3009104, INCB028050) 1187594-09-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062524
Encompass_Preferred
Baricitinib (LY3009104 INCB028050 CAS 1187594-09-7) is an orally bioavailable ATP-competitive inhibitor targeting Janus kinases JAK1 and JAK2 It also exhibits moderate inhibition of TYK2 and weaker inhibition against JAK3 In biochemical assays baricitinib inhibits JAK1 and JAK2 with IC50 values of 5 9 nM and 5 7 nM respectively At concentrations below 50 nM baricitinib suppresses intracellular signaling mediated by several pro-inflammatory cytokines notably IL-6 and IL-23 Due to its mechanistic profile baricitinib is actively explored in inflammatory disease research such as rheumatoid arthritis
Catalog No. 50-000-62524 Supplier Apexbio Technology LLC Supplier No. A41415.1
Baricitinib (LY3009104 INCB028050 CAS 1187594-09-7) is an orally bioavailable ATP-competitive inhibitor targeting Janus kinases JAK1 and JAK2 It also exhibits moderate inhibition of TYK2 and weaker inhibition against JAK3 In biochemical assays baricitinib inhibits JAK1 and JAK2 with IC50 values of 5 9 nM and 5 7 nM respectively At concentrations below 50 nM baricitinib suppresses intracellular signaling mediated by several pro-inflammatory cytokines notably IL-6 and IL-23 Due to its mechanistic profile baricitinib is actively explored in inflammatory disease research such as rheumatoid arthritis