Learn More
Cayman Chemical Baricitinib, Purity: ≥95%, CAS: 1187594-09-7, 50 mg
SDP
Catalog No. 502968279
Encompass_Preferred
A JAK1 and JAK2 inhibitor (IC50s = 0.0059 and 0.0057 µM, respectively); selective for JAK1 and JAK2 over JAK3, TYK2, c-Met, and Chk2 (IC50s = >0.4, 0.053, >10, and >1 µM, respectively); inhibits IL-6-induced phosphorylation of STAT3 in isolated human PBMCs and rat whole blood (IC50s = 0.044 and 0.128 µM, respectively); reduces disease severity in a type II collagen-induced mouse model of arthritis at 3 and 10 mg/kg; decreases lung macrophage infiltration and disease severity in rhesus monkeys infected with SARS-CoV-2 at 4 mg/animal
Catalog No. 50-296-8279 Supplier Cayman Chemical Supplier No. 1670750 MG
A JAK1 and JAK2 inhibitor (IC50s = 0.0059 and 0.0057 µM, respectively); selective for JAK1 and JAK2 over JAK3, TYK2, c-Met, and Chk2 (IC50s = >0.4, 0.053, >10, and >1 µM, respectively); inhibits IL-6-induced phosphorylation of STAT3 in isolated human PBMCs and rat whole blood (IC50s = 0.044 and 0.128 µM, respectively); reduces disease severity in a type II collagen-induced mouse model of arthritis at 3 and 10 mg/kg; decreases lung macrophage infiltration and disease severity in rhesus monkeys infected with SARS-CoV-2 at 4 mg/animal