Learn More
AdipoGen Baricitinib (50 mg)
SDP
Supplier:  AdipoGen AGCR13734M050
Encompass_Preferred
Chemical. CAS: 1187594-09-7. Formula: C16H17N7O2S. MW: 371.4. Baricitinib is a selective orally bioavailable JAK1/JAK2 inhibitor with nanomolar potency against JAK1 (IC50 = 5.9nM) and JAK2 (IC50 = 5.7nM) and inhibits Tyk2 (IC50 = 53nM). It displays >100-fold selectivity for JAK1/2 over JAK3 (IC50 > 400nM). Janus kinases (JAKs) are non-receptor kinases and play important roles in the proinflammatory signaling pathways that are frequently over-activated in autoimmune disorders such as rheumatoid arthritis. Upon binding of extracellular cytokines and growth factors, JAKs are phosphorylated and activate signal transducers and activators of transcription (STATs). Via these signaling cascades, inflammatory cytokine and chemokine transcription is induced to form inflammatory mediators including IL-2, IL-6, IL-12, IL-15, IL-23. In addition the drug’s anti-inflammatory activity is expected to act on the inflammatory cascade associated with COVID-19.
Catalog No. 50-205-4153