Learn More
Selleck Chemical LLC Baricitinib (INCB028050) 5mg 1187594-09-7 LY3009104
SDP
Supplier:  Selleck Chemical LLC S28515MG
Encompass_Preferred
Baricitinib (LY3009104, INCB028050) is a selective JAK1 and JAK2 inhibitor with IC50 of 5.9 nM and 5.7 nM in cell-free assays, ~70 and ~10-fold selective versus JAK3 and Tyk2, no inhibition to c-Met and Chk2. Baricitinib is found to reduce or interrupt the passage of the virus into target cells and is used in the treatment research for COVID-19. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6227