Learn More
Medchemexpress LLC Baricitinib-d3 | 1564242-30-3 | C16H14D3N7O2S | 1 MG
SDP
Catalog No. 5000374031
Encompass_Preferred
Baricitinib-d3 is the deuterium labeled version of Baricitinib, a selective and orally bioavailable JAK1 and JAK2 inhibitor with IC50s of 5.9 nM and 5.7 nM, respectively. It is intended for research use only.
  • Deuterium labeled Baricitinib
  • Selective and orally bioavailable JAK1 and JAK2 inhibitor
  • Suitable as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-74031 Supplier Medchemexpress LLC Supplier No. HY15315S11MG
Baricitinib-d3 is the deuterium labeled version of Baricitinib, a selective and orally bioavailable JAK1 and JAK2 inhibitor with IC50s of 5.9 nM and 5.7 nM, respectively. It is intended for research use only.
  • Deuterium labeled Baricitinib
  • Selective and orally bioavailable JAK1 and JAK2 inhibitor
  • Suitable as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs