Learn More
Medchemexpress LLC Baricitinib phosphate | 1187595-84-1 | C16H20N7O6PS | 5 MG
SDP
Catalog No. 5000372927
Encompass_Preferred
Baricitinib phosphate is a selective, orally bioavailable JAK1/JAK2 inhibitor with IC50 values of 5.9 nM and 5.7 nM, respectively. It is intended for research use only.
  • Selective orally bioavailable JAK1/JAK2 inhibitor.
  • Potent inhibitor of JAK signaling and function in cell-based assays.
  • Inhibits IL-6-stimulated STAT3 phosphorylation and MCP-1 production in PBMCs.
  • Inhibits IL-23-stimulated pSTAT3 in naive T-cells, preventing IL-17 and IL-22 production.
  • Demonstrates in vivo efficacy in reducing inflammation and paw swelling.
  • Reduces CD8 infiltration and MHC class I and class II expression in animal models.
Catalog No. 50-003-72927 Supplier Medchemexpress LLC Supplier No. HY15315A5MG
Baricitinib phosphate is a selective, orally bioavailable JAK1/JAK2 inhibitor with IC50 values of 5.9 nM and 5.7 nM, respectively. It is intended for research use only.
  • Selective orally bioavailable JAK1/JAK2 inhibitor.
  • Potent inhibitor of JAK signaling and function in cell-based assays.
  • Inhibits IL-6-stimulated STAT3 phosphorylation and MCP-1 production in PBMCs.
  • Inhibits IL-23-stimulated pSTAT3 in naive T-cells, preventing IL-17 and IL-22 production.
  • Demonstrates in vivo efficacy in reducing inflammation and paw swelling.
  • Reduces CD8 infiltration and MHC class I and class II expression in animal models.