Learn More
Medchemexpress LLC Baricitinib phosphate | 1187595-84-1 | C16H20N7O6PS | 500 MG
SDP
Catalog No. 5000373155
Encompass_Preferred
Baricitinib phosphate is a selective, orally bioavailable inhibitor targeting JAK1 and JAK2 pathways. It demonstrates potent inhibition of JAK signaling and function in various cellular assays, making it suitable for research applications.
  • Selective JAK1/JAK2 inhibitor with high potency
  • Inhibits IL-6-stimulated STAT3 phosphorylation and MCP-1 production in PBMCs
  • Inhibits pSTAT3 stimulated by IL-23 in isolated naive T-cells
  • Prevents IL-17 and IL-22 production by Th17 cells
  • Demonstrates anti-inflammatory effects in vivo, reducing hind paw swelling and immune cell infiltration
Catalog No. 50-003-73155 Supplier Medchemexpress LLC Supplier No. HY15315A500MG
Baricitinib phosphate is a selective, orally bioavailable inhibitor targeting JAK1 and JAK2 pathways. It demonstrates potent inhibition of JAK signaling and function in various cellular assays, making it suitable for research applications.
  • Selective JAK1/JAK2 inhibitor with high potency
  • Inhibits IL-6-stimulated STAT3 phosphorylation and MCP-1 production in PBMCs
  • Inhibits pSTAT3 stimulated by IL-23 in isolated naive T-cells
  • Prevents IL-17 and IL-22 production by Th17 cells
  • Demonstrates anti-inflammatory effects in vivo, reducing hind paw swelling and immune cell infiltration