Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Baricitinib phosphate | 1187595-84-1 | MFCD25976707 | 100.0% | 469.41 g/mol | C16H20N7O6PS | 10 MG
SDP

Catalog No. 5000286216
Encompass_Preferred

Baricitinib phosphate is the phosphate salt form of baricitinib, a selective, orally bioavailable inhibitor of JAK1 and JAK2 with low-nanomolar potency. It is supplied as a solid research reagent intended for in vitro and in vivo studies probing JAK-mediated signaling.

  • Selective inhibitor of JAK1 and JAK2 (IC50: JAK1 5.9 nM; JAK2 5.7 nM; TYK2 53 nM; JAK3 560 nM).
  • Molecular formula C16H20N7O6PS and molecular weight 469.41 g/mol.
  • CAS number 1187595-84-1.
  • Purity 99.97% as listed by manufacturer.
  • High solubility in DMSO (250 mg/mL; 532.58 mM); in vivo formulations show solubility ≥2.08 mg/mL (4.43 mM).
  • Appearance white to yellow solid suitable for research use.
  • Store sealed at 4°C; in solvent store at -80°C (1 year) or -20°C (6 months).

Catalog No. 50-002-86216 Supplier Medchemexpress LLC Supplier No. HY15315A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Baricitinib phosphate is the phosphate salt form of baricitinib, a selective, orally bioavailable inhibitor of JAK1 and JAK2 with low-nanomolar potency. It is supplied as a solid research reagent intended for in vitro and in vivo studies probing JAK-mediated signaling.

  • Selective inhibitor of JAK1 and JAK2 (IC50: JAK1 5.9 nM; JAK2 5.7 nM; TYK2 53 nM; JAK3 560 nM).
  • Molecular formula C16H20N7O6PS and molecular weight 469.41 g/mol.
  • CAS number 1187595-84-1.
  • Purity 99.97% as listed by manufacturer.
  • High solubility in DMSO (250 mg/mL; 532.58 mM); in vivo formulations show solubility ≥2.08 mg/mL (4.43 mM).
  • Appearance white to yellow solid suitable for research use.
  • Store sealed at 4°C; in solvent store at -80°C (1 year) or -20°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.